missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) oxide, 97%

Product Code. 15463087
Change view
Click to view available options
Quantity:
500 g
2500 g
Unit Size:
2500g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15463087 2500 g 2500g
15473087 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15463087 Supplier Thermo Scientific Alfa Aesar Supplier No. A14436.0E
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1317-39-1 | Cu2O

It is used as a catalyst in decarboxylation reactions of arylacetic acids and of malonic acids under mild conditions. It is also used in batteries and electrodes, ceramic colorant, catalyst, purification of hydrogen, reagent in analytical chemistry, metallurgical and welding fluxes. It finds its application also as an antifouling agent for marine paints and in Benedict's Test. It is used to provide a source supplemental elemental copper.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a catalyst in decarboxylation reactions of arylacetic acids and of malonic acids under mild conditions. It is also used in batteries and electrodes, ceramic colorant, catalyst, purification of hydrogen, reagent in analytical chemistry, metallurgical and welding fluxes. It finds its application also as an antifouling agent for marine paints and in Benedict′s Test. It is used to provide a source supplemental elemental copper.

Solubility
Insoluble in water. Dissolves in concentrated ammonia.

Notes
Air and moisture sensitive. Store away from air, water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1317-39-1
Molecular Formula Cu2O
MDL Number MFCD00010974
Synonym Cuprous oxide

Specifications

Boiling Point 1800°C (decomposition)
Melting Point 1235°C
Quantity 2500 g
UN Number UN3077
Merck Index 14,2664
Solubility Information Insoluble in water. Dissolves in concentrated ammonia.
Formula Weight 143.08
Percent Purity 97%
Odor Odorless
Sensitivity Air and moisture sensitive
Density 6
Chemical Name or Material Copper(I) oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.