missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) Oxide, 97%, Solstice

Product Code. 15641870 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 kg
12 kg
Packaging:
Poly Bottle
Unit Size:
12kg
25g
2kg
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15641870 25 g Poly Bottle 25g
15661870 500 g Poly Bottle 500g
15651870 2 kg Poly Bottle 2kg
15631870 12 kg Poly Bottle 12kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15641870 Supplier Honeywell-Fluka Supplier No. 20882525G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Powder, <5 μm, 97%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1317-39-1
MDL Number MFCD00010974

Specifications

Melting Point 1230°C
Color Brown or Red
pH 8.0 (at 100g/L 20°C)
Physical Form Powder
Quantity 25 g
Linear Formula Cu2O
UN Number UN3077
Formula Weight 143.09g/mol
Percent Purity 97%
Packaging Poly Bottle
Density 6g/mL at 25°C
Chemical Name or Material Copper(I) Oxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.