missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) Oxide, 97%, Honeywell Fluka™

Product Code. 15651870 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
25 g
500 g
2 kg
12 kg
Packaging:
Poly Bottle
Unit Size:
12kg
25g
2kg
500g
This item is not returnable. View return policy

Product Code. 15651870

Brand: Honeywell-Fluka 2088252KG

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Powder, <5 μm, 97%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1317-39-1
MDL Number MFCD00010974

Specifications

Melting Point 1230°C
Color Brown or Red
pH 8.0 (at 100g/L 20°C)
Physical Form Powder
Quantity 2 kg
Linear Formula Cu2O
UN Number UN3077
Formula Weight 143.09g/mol
Percent Purity 97%
Packaging Poly Bottle
Density 6g/mL at 25°C
Chemical Name or Material Copper(I) Oxide
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.