missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) oxide, 97%

Product Code. 10318803 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
2.5kg
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10318803 2.5 kg Glass bottle 2.5kg
10328480 25 g Glass bottle 25g
10432011 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10318803 Supplier Thermo Scientific Acros Supplier No. 197420025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1317-39-1 | Cu2O | 143.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1317-39-1
Molecular Formula Cu2O
Molecular Weight (g/mol) 143.09
MDL Number MFCD00010974
Synonym Cuprous oxide

Specifications

Boiling Point 1800.0°C
Melting Point 1232.0°C
Color Brown-Red
Physical Form Powder
Quantity 2.5 kg
Assay Percent Range 96% min. (Complexometry)
Linear Formula Cu2O
Merck Index 15, 2654
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ammonia or min.acids
Formula Weight 143.09
Percent Purity 97%
Packaging Glass bottle
Chemical Name or Material Copper(I) oxide, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.