missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(I) iodide, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
29.40 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Copper(I) iodide is used as a catalyst in organic synthesis, cathode ray tube coating, cloud seeding, and as temperature indicator with HgI2. It catalyzes the conversion of aryl, heteroaryl, and vinyl bromides into the corresponding iodides, which are employed in coupling reactions such as Suzuki, Stille, Sonogashira, Heck and Ullmann reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | ∼1290°C |
Melting Point | 605°C |
Color | Brown to White |
Physical Form | Chunks and/or Powder |
Quantity | 5 g |
Assay Percent Range | 99.998% (Metal basis) |
Solubility Information | Copper(I) iodide is soluble in ammonia,thiosulfate,sodium and potassium iodide solutions. Insoluble in water and dilute acids. |
Formula Weight | 190.45 |
Odor | Weak odor |
Refractive Index | 2.346 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'