missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) iodide, 99.995%, (trace metal basis)

Product Code. 10642983 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10642983

Brand: Thermo Scientific Acros 201501000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7681-65-4 | CuI | 190.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent, for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-65-4
Molecular Formula CuI
Molecular Weight (g/mol) 190.45
MDL Number MFCD00010978
Synonym cuprous iodide, copper l iodide, copper i iodide metals basis, marshite, copper i iodide 250g

Specifications

Boiling Point 1290°C
Melting Point 605°C
Color Brown or Gray to White
Physical Form Powder
Quantity 100 g
Assay Percent Range 31.0 to 34% (Cu) (Complexometry)
Linear Formula CuI
Fieser 01,169; 02,92; 03,69; 05,167; 06,147; 07,81; 08,121; 09,124; 10,107; 11,141; 12,141; 16,98; 17,88
Merck Index 15, 2652
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ammonia and potassium iodide solutions,practically insoluble in alcohol
Formula Weight 190.45
Percent Purity 100.00%
Grade Trace Metal Basis
Packaging Glass bottle
Density 5.62
Chemical Name or Material Copper(I) iodide, 99.995%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Copper(I) iodide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.