missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) Chloride, TRC

Product Code. 30353895
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30353895 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30353895 Supplier Toronto Research Chemicals Supplier No. TRCC68528050G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material Copper(I) Chloride
CAS 7758-89-6
Molecular Formula ClCu
Quantity 50 g
Synonym Copper Chloride (CuCl), Copper Chloride (Cu2Cl2), Copper Monochloride, Copper(1+) Chloride, Cuprous Chloride, Cuprous Chloride (Cu2Cl2), Cuprous Chloride (CuCl), Dicopper Dichloride
InChI Formula InChI=1S/ClH.Cu/h1H;/q;+1/p-1
SMILES Cl[Cu]
IUPAC Name chlorocopper
Molecular Weight (g/mol) 99
Formula Weight 97.8985
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.