missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(I) Chloride, ≥97.0% (RT), Honeywell Fluka™
Click to view available options
Quantity:
25 g
250 g
1 kg
Packaging:
Poly bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Boiling Point | 1345°C (1013hPa) |
Melting Point | 492°C |
Color | Colorless |
Physical Form | Powder |
Quantity | 1 kg |
Linear Formula | CuBr |
UN Number | NONH for all modes of transport |
Formula Weight | 143.45g/mol |
Percent Purity | ≥98.0% (RT) |
Odor | Odorless |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'