Learn More
Copper(I) bromide, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
Description
It is used as effective catalyst for tetrahydropyranylation of alcohols. It can also be used in combination with palladium in catalytic synthesis of 3-haloindoles through annulation process. It is widely used in the synthesis of organic fine chemicals. The compound is widely used in the synthesis of organic compounds. It is also useful for copper-mediated coupling reactions. A silica gel support copper bromide hexa methyltriethylenetetramine complex has shown useful application for the atom transfer radical polymerization of methyl methacrylate in toluene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 1345°C |
Melting Point | 492°C |
Color | Green |
Physical Form | Powder |
Quantity | 5 g |
Assay Percent Range | 99.998% (Metal basis) |
Solubility Information | Soluble in hydrogen bromide,hydrochloric acid,and ammonium hydroxide. Slightly soluble in water. Insoluble in acetone. |
Formula Weight | 143.45 |
Sensitivity | Air Sensitive |
Appearance | Light green powder |
Show More |
RUO – Research Use Only