missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) Bromide, ≥98.0% (RT), Honeywell™

Product Code. 15696010 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
Packaging:
Poly bottle
Unit Size:
100g
10g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15696010 10 g Poly bottle 10g
15686010 100 g Poly bottle 100g
15606020 500 g Poly bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15696010 Supplier Honeywell Research Chemicals Supplier No. 6116310G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Purum p.a.,≥98.0% (RT)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7787-70-4
Molecular Formula BrCu
Molecular Weight (g/mol) 143.45
MDL Number MFCD00010969
InChI Key NKNDPYCGAZPOFS-UHFFFAOYSA-M
Synonym copper i bromide, copper bromide cubr, copper 1+ bromide, copper monobromide, cuprousbromide, hsdb 270, cubr, copper 1 bromide
PubChem CID 24593
SMILES [Cu+].[Br-]

Specifications

Boiling Point 1345°C (1013hPa)
Melting Point 492°C
Color Colorless
Physical Form Powder
Quantity 10 g
Linear Formula CuBr
UN Number NONH for all modes of transport
IUPAC Name λ¹-copper(1+) bromide
Formula Weight 143.45g/mol
Percent Purity ≥98.0% (RT)
Odor Odorless
Packaging Poly bottle
Density 4.71g/mL at 25°C
Chemical Name or Material Copper(I) bromide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.