missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, High Leaded Tin Bronze, CDA 941, C94100, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Copper, High Leaded Tin Bronze, CDA 941, C94100, Ø38x19mm
Specifications
Specifications
| Chemical Name or Material | Aluminium/Manganese/Silicon/Iron/Phosphorus/Arsenic/Bismuth/Antimony/Nickel/Zinc/Tin/Lead/Chromium/Cobalt/Carbon/Silver/Copper |
| CAS | 7440-50-8 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Aluminium 0.0016 %/Manganese 0.002 %/Silicon 0.002 %/Iron 0.003 %/Phosphorus 0.008 %/Arsenic 0.01 %/Bismuth 0.03 %/Antimony 0.27 %/Nickel 0.3 %/Zinc 0.37 %/Tin 6 %/Lead 19 %/Chromium (0.001) %/Cobalt (0.001) %/Carbon (0.004) %/Silver (0.01) %/Copper (74) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction