Learn More
Thermo Scientific Alfa Aesar COPPER FOIL 0.025MM 99.8% 30X30CM
Brand: Thermo Scientific Alfa Aesar 013382.RF
COPPER FOIL 0.025MM 99.8% 30X30CM
Code : NEW
Additional Details : CAS Number : 7440-50-8 Weight : 0.01000kg
Brand: Thermo Scientific Alfa Aesar 013382.RF
COPPER FOIL 0.025MM 99.8% 30X30CM
Code : NEW
Additional Details : CAS Number : 7440-50-8 Weight : 0.01000kg