missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Copper Nickel, 50g, ARMI∣MBH
Click to view available options
Quantity:
50 g
Unit Size:
50g
Description
Copper, Copper Nickel, 50g
Specifications
Specifications
| Chemical Name or Material | Aluminium/Carbon/Chromium/Niobium/Tin/Magnesium/Titanium/Iron/Phosphorus/Lead/Silicon/Cobalt/Manganese/Zinc/Nickel/Copper |
| CAS | 7440-50-8 |
| Quantity | 50 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Aluminium (0.001) %/Carbon (0.0013) %/Chromium (0.004) %/Niobium (0.02) %/Tin (0.025) %/Magnesium (0.03) %/Titanium (0.036) %/Iron (0.04) %/Phosphorus (0.04) %/Lead (0.046) %/Silicon (0.05) %/Cobalt (0.195) %/Manganese (0.7) %/Zinc (1.02) %/Nickel (11.4) %/Copper (86.3) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction