missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Copper Nickel, 50g, ARMI∣MBH
Click to view available options
Quantity:
50 g
Unit Size:
50g
Description
Copper, Copper Nickel, 50g
Specifications
Specifications
| Chemical Name or Material | Beryllium/Boron/Tin/Magnesium/Lead/Titanium/Cobalt/Phosphorus/Chromium/Zinc/Manganese/Iron/Niobium/Silicon/Nickel/Copper/Carbon |
| CAS | 7440-50-8 |
| Quantity | 50 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Beryllium 0.0035 %/Boron 0.0053 %/Tin 0.009 %/Magnesium 0.0106 %/Lead 0.012 %/Titanium 0.0203 %/Cobalt 0.0238 %/Phosphorus 0.034 %/Chromium 0.141 %/Zinc 0.209 %/Manganese 0.217 %/Iron 0.347 %/Niobium 0.43 %/Silicon 0.689 %/Nickel 31.03 %/Copper 66.67 %/Carbon (0.008) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction