missing translation for 'onlineSavingsMsg'
Learn More

Copper Citrate, TRC

Product Code. 30349946
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30349946 5 g -
30366540 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30349946 Supplier Toronto Research Chemicals Supplier No. TRCC6853005G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10402-15-0
Molecular Formula C6H8O7 xCu
Molecular Weight (g/mol) 255.67
Synonym 2-Hydroxy-1,2,3-propanetricarboxylic Acid Copper Salt, Citric Acid Copper Salt
IUPAC Name copper;2-oxidopropane-1,2,3-tricarboxylate
SMILES O.O=C1CC2(CC(=O)O[Cu]O2)C(=O)O[Cu]O1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C6H7O7.Cu/c7-3(8)1-6(13,5(11)12)2-4(9)10;/h1-2H2,(H,7,8)(H,9,10)(H,11,12);/q-1;+2/p-3
Formula Weight 250.9264
Chemical Name or Material Copper Citrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.