missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Brass Traces, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Copper, Brass Traces, Ø40x15mm
Specifications
Specifications
| Chemical Name or Material | Boron/Tellurium/Cobalt/Bismuth/Cadmium/Silver/Iron/Silicon/Antimony/Nickel/Manganese/Aluminium/Arsenic/Lead/Tin/Zinc/Copper |
| CAS | 7440-50-8 |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Boron 0.0015 %/Tellurium 0.003 %/Cobalt 0.004 %/Bismuth 0.008 %/Cadmium 0.01 %/Silver 0.013 %/Iron 0.02 %/Silicon 0.02 %/Antimony 0.03 %/Nickel 0.03 %/Manganese 0.035 %/Aluminium 0.04 %/Arsenic 0.048 %/Lead 0.12 %/Tin 0.14 %/Zinc 37 %/Copper [bal] % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction