missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Brass Traces, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Copper, Brass Traces, Ø40x15mm
Specifications
Specifications
| Chemical Name or Material | Chromium/Cobalt/Aluminium/Iron/Nickel/Silicon/Tin/Silver/Antimony/Manganese/Bismuth/Arsenic/Cadmium/Lead/Zinc/Copper |
| CAS | 7440-50-8 |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Chromium (0.003) %/Cobalt (0.02) %/Aluminium (0.07) %/Iron (0.097) %/Nickel (0.105) %/Silicon (0.112) %/Tin (0.133) %/Silver (0.22) %/Antimony (0.23) %/Manganese (0.285) %/Bismuth (0.29) %/Arsenic (0.39) %/Cadmium (0.47) %/Lead (0.57) %/Zinc (35.6) %/Copper (61.5) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction