missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Brass Traces, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Copper, Brass Traces, Ø40x15mm
Specifications
Specifications
| Chemical Name or Material | Cadmium/Chromium/Cobalt/Silver/Arsenic/Bismuth/Tellurium/Aluminium/Antimony/Silicon/Iron/Lead/Nickel/Tin/Manganese/Zinc/Copper |
| CAS | 7440-50-8 |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Cadmium (0.0014) %/Chromium (0.0055) %/Cobalt (0.015) %/Silver (0.015) %/Arsenic (0.017) %/Bismuth (0.017) %/Tellurium (0.022) %/Aluminium (0.03) %/Antimony (0.05) %/Silicon (0.05) %/Iron (0.058) %/Lead (0.067) %/Nickel (0.076) %/Tin (0.13) %/Manganese (0.145) %/Zinc (35.9) %/Copper (63.8) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction