missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Brass Traces, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Copper, Brass Traces, Ø40x15mm
Specifications
Specifications
| Chemical Name or Material | Boron/Selenium/Cobalt/Chromium/Phosphorus/Cadmium/Bismuth/Arsenic/Silver/Iron/Silicon/Antimony/Aluminium/Nickel/Tin/Manganese/Lead/Zinc/Copper/Zirconium/Sulfur/Tellurium |
| CAS | 7440-50-8 |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Boron 0.0005 %/Selenium 0.0008 %/Cobalt 0.0017 %/Chromium 0.0024 %/Phosphorus 0.008 %/Cadmium 0.0122 %/Bismuth 0.049 %/Arsenic 0.052 %/Silver 0.061 %/Iron 0.072 %/Silicon 0.097 %/Antimony 0.101 %/Aluminium 0.174 %/Nickel 0.199 %/Tin 0.214 %/Manganese 0.287 %/Lead 0.325 %/Zinc 36.8 %/Copper 61.5 %/Zirconium (0.001) %/Sulfur (0.002) %/Tellurium (0.004) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction