missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Brass Traces, 50g, ARMI∣MBH
Click to view available options
Quantity:
50 g
Unit Size:
50g
Description
Copper, Brass Traces, 50g
Specifications
Specifications
| Chemical Name or Material | Manganese/Cadmium/Tellurium/Aluminium/Bismuth/Cobalt/Antimony/Arsenic/Nickel/Tin/Silicon/Lead/Iron/Zinc/Copper/Boron |
| CAS | 7440-50-8 |
| Quantity | 50 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Manganese 0.0013 %/Cadmium 0.0032 %/Tellurium 0.0035 %/Aluminium 0.0041 %/Bismuth 0.0058 %/Cobalt 0.0067 %/Antimony 0.0095 %/Arsenic 0.0106 %/Nickel 0.0133 %/Tin 0.0197 %/Silicon 0.0203 %/Lead 0.0246 %/Iron 0.034 %/Zinc 33.64 %/Copper 66.07 %/Boron (0.0004) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction