missing translation for 'onlineSavingsMsg'
Learn More

Copper Beryllium foil, alloy C17200, 0.4mm (0.016in) thick

Product Code. 15436415
Click to view available options
Quantity:
50 x 200 mm
Unit Size:
Each
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 15436415

Brand: Thermo Scientific Alfa Aesar 041914.GU

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

 

Uses include springs of all kinds where high strength and corrosion resistance are important, resistance welding electrodes, dies and high strength, corrosion resistant tubing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Copper Beryllium foil, Alloy C17200
Name Note 0.4mm (0.016 in.) thick
Avg. Mol. Wt. or Mol. Wt. Range Cu:Be; 98.1:1.9 wt%
Quantity 50 x 200 mm
Odor Odorless
Weight ≈57.4g/75x225mm
Form Foil
MDL Number MFCD01320550
Solubility Information Insoluble in water.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.