missing translation for 'onlineSavingsMsg'
Learn More

Copper based methanol synthesis catalyst, Thermo Scientific Chemicals

Product Code. 15442437
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
Unit Size:
1kg
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15442437 500 g 500g
15452437 1 kg 1kg
15462437 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15442437 Supplier Thermo Scientific Alfa Aesar Supplier No. 045776.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Lurgi-low pressure process for methanol synthesis also uses a copper based catalyst which is CuO/ZnO.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Color Brown to Black
Quantity 500 g
UN Number UN3077
Solubility Information Insoluble in water.
Physical Form Pellets
Chemical Name or Material Copper based methanol synthesis catalyst

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.