missing translation for 'onlineSavingsMsg'
Learn More

Copper aluminum oxide, 99.5% (metals basis)

Product Code. 11388198
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11388198 250 g 250g
11378198 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11388198 Supplier Thermo Scientific Alfa Aesar Supplier No. 039419.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12042-92-1 | Al2CuO4 | 181.51 g/mol

Applications for copper aluminum oxide nanocrystalsare included in microbatteries, in micro-electronics, circuits and in nanowire, and in nanofluids.It is also a promising material for thermoelectric devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12042-92-1
Molecular Formula Al2CuO4
Molecular Weight (g/mol) 181.51
MDL Number MFCD00210610
InChI Key IPJRADJOSNLGBF-UHFFFAOYSA-N
PubChem CID 12947132
IUPAC Name copper;oxido(oxo)alumane
SMILES [Cu++].[O-][Al]=O.[O-][Al]=O

Specifications

Physical Form Powder
Quantity 250 g
Solubility Information Insoluble in water.
Formula Weight 181.5
Percent Purity 99.5%
Assay (metals basis)
Chemical Name or Material Copper aluminum oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.