missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Aluminum Bronze, CDA 954M, C95410, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Copper, Aluminum Bronze, CDA 954M, C95410, Ø38x3mm
Specifications
Specifications
| Chemical Name or Material | Lead/Tin/Carbon/Phosphorus/Chromium/Cobalt/Silver/Silicon/Manganese/Zinc/Nickel/Iron/Aluminium/Copper/Antimony/Arsenic |
| CAS | 7440-50-8 |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Lead 0.004 %/Tin 0.005 %/Carbon 0.006 %/Phosphorus 0.007 %/Chromium 0.008 %/Cobalt 0.008 %/Silver 0.009 %/Silicon 0.034 %/Manganese 0.052 %/Zinc 0.22 %/Nickel 1.95 %/Iron 3.87 %/Aluminium 10.55 %/Copper 83.3 %/Antimony <0.01 %/Arsenic <0.01 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction