missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Tin Zinc Bismuth, Federalloy III-932, C89835, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Copper, Tin Zinc Bismuth, Federalloy III-932, C89835, Ø38x3mm
Specifications
Specifications
| Chemical Name or Material | Selenium/Silver/Aluminium/Manganese/Silicon/Arsenic/Antimony/Lead/Phosphorus/Iron/Nickel/Bismuth/Zinc/Tin/Cobalt/Carbon/Chromium/Copper |
| CAS | 7440-50-8 |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Selenium 0.001 %/Silver 0.001 %/Aluminium 0.002 %/Manganese 0.002 %/Silicon 0.002 %/Arsenic 0.004 %/Antimony 0.01 %/Lead 0.01 %/Phosphorus 0.032 %/Iron 0.035 %/Nickel 0.46 %/Bismuth 2.37 %/Zinc 3.48 %/Tin 6.9 %/Cobalt (0.001) %/Carbon (0.002) %/Chromium (0.002) %/Copper (87) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction