missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Copper Nickel 10%, CDA 706, C70600, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Copper, Copper Nickel 10%, CDA 706, C70600, Ø38x3mm
Specifications
Specifications
| Chemical Name or Material | Antimony/Silver/Carbon/Silicon/Cobalt/Manganese/Lead/Phosphorus/Tin/Zinc/Iron/Nickel/Copper/Beryllium/Bismuth/Cadmium/Tellurium/Zirconium/Magnesium/Chromium/Aluminium/Arsenic/Selenium/Oxygen |
| CAS | 7440-50-8 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Antimony 0.0012 %/Silver 0.002 %/Carbon 0.003 %/Silicon 0.004 %/Cobalt 0.006 %/Manganese 0.04 %/Lead 0.047 %/Phosphorus 0.06 %/Tin 0.13 %/Zinc 0.13 %/Iron 0.28 %/Nickel 9.8 %/Copper 89.4 %/Beryllium (<0.002) %/Bismuth (<0.005) %/Cadmium (<0.02) %/Tellurium (<0.03) %/Zirconium (0.0004) %/Magnesium (0.0007) %/Chromium (0.0008) %/Aluminium (0.002) %/Arsenic (0.002) %/Selenium (0.002) %/Oxygen (0.008) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction