missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Free Cutting Brass, CDA 360, C36000, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
Each
Description
Copper, Free Cutting Brass, CDA 360, C36000, 100g
Specifications
Specifications
| Chemical Name or Material | Cobalt/Bismuth/Carbon/Chromium/Phosphorus/Cadmium/Aluminium/Silicon/Silver/Antimony/Manganese/Arsenic/Nickel/Iron/Tin/Lead/Zinc/Copper/Boron/Hydrogen/Magnesium/Molybdenum/Niobium/Nitrogen/Oxygen/Palladium/Selenium/Sulfur/Tellurium/Titanium/Vanadium |
| CAS | 7440-50-8 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Cobalt 0.001 %/Bismuth 0.0023 %/Carbon 0.003 %/Chromium 0.003 %/Phosphorus 0.003 %/Cadmium 0.0034 %/Aluminium 0.01 %/Silicon 0.01 %/Silver 0.011 %/Antimony 0.012 %/Manganese 0.0131 %/Arsenic 0.026 %/Nickel 0.12 %/Iron 0.27 %/Tin 0.29 %/Lead 2.73 %/Zinc 35.1 %/Copper 61.6 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction