missing translation for 'onlineSavingsMsg'
Learn More

Copper, 99.9998%, (trace metal basis), Thermo Scientific™

Product Code. 10555090
Click to view available options
Quantity:
25 g
125 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-50-8
Molecular Formula Cu
Molecular Weight (g/mol) 63.55
MDL Number MFCD00010965
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
Synonym cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
PubChem CID 23978
ChEBI CHEBI:30052
IUPAC Name copper
SMILES [Cu]

Specifications

Color Brown-Rust
Melting Point 1083.0°C
Boiling Point 2595.0°C
Assay Percent Range 99.9998%
Linear Formula Cu
Quantity 25 g
Solubility Information Solubility in water: insoluble.
Formula Weight 63.54
Percent Purity 99.9998%
Grade Trace Metal Basis
Packaging Glass bottle
Form Shot
Chemical Name or Material Copper
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'