missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COPG Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565194
This item is not returnable.
View return policy
Description
Immunogen sequence: MLPSILVLLKR CVMDDDNEVR DRATFYLNVL EQKQKALNAG YILNGLTVSI PGLERALQQY TLEPSEKPFD LKSVPL Highest antigen sequence identity to the following orthologs - mouse 99%, rat 99%.
The coatomer is a cytosolic protein complex that binds to dilysine motifs and reversibly associates with Golgi non-clathrin-coated vesicles, which further mediate biosynthetic protein transport from the ER, via the Golgi up to the trans Golgi network. Coatomer complex is required for budding from Golgi membranes, and is essential for the retrograde Golgi-to-ER transport of dilysine-tagged proteins. In mammals, the coatomer can only be recruited by membranes associated to ADP-ribosylation factors (ARFs), which are small GTP-binding proteins; the complex also influences the Golgi structural integrity, as well as the processing, activity, and endocytic recycling of LDL receptors. Required for limiting lipid storage in lipid droplets. Involved in lipid homeostasis by regulating the presence of perilipin family members PLIN2 and PLIN3 at the lipid droplet surface and promoting the association of adipocyte triglyceride lipase (PNPLA2) with the lipid droplet surface to mediate lipolysis. [UniProt].
Specifications
| COPG | |
| Polyclonal | |
| Unconjugated | |
| Copg1 | |
| AU019265; BC056168; coat protein gamma-cop; coatomer protein complex subunit gamma 1; coatomer protein complex, subunit gamma 1; coatomer subunit gamma; coatomer subunit gamma-1; COPG; COPG1; D6Ertd71e; D6Wsu16e; Gamma-1-coat protein; gamma-1-COP; gamma-coat protein; gamma-COP | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22820 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y678 | |
| Copg1 | |
| Recombinant Human COPG. Recombinant protein control fragment (Product #RP-104681). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction