missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
COPG Polyclonal specifically detects COPG in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | COPG |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 350 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate |
| Gene Alias | coat protein gamma-cop, coatomer protein complex, subunit gamma, coatomer protein complex, subunit gamma 1, COPG1coatomer subunit gamma, FLJ21068, Gamma-coat protein, gamma-COP |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human COPG. |
| Purification Method | Antigen Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?