missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COPE Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559645
This item is not returnable.
View return policy
Description
Immunogen sequence: DATLTQLATA WVSLATGGEK LQDAYYIFQE MADKCSPTLL LLNGQAACHM AQGRWEAAEG LLQEALDKDS GYPETLVNL Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.
The product of this gene is an epsilon subunit of coatomer protein complex. Coatomer is a cytosolic protein complex that binds to dilysine motifs and reversibly associates with Golgi non-clathrin-coated vesicles. It is required for budding from Golgi membranes, and is essential for the retrograde Golgi-to-ER transport of dilysine-tagged proteins. Coatomer complex consists of at least the alpha, beta, beta', gamma, delta, epsilon and zeta subunits. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
| COPE | |
| Polyclonal | |
| Unconjugated | |
| COPE | |
| 1110005D17Rik; coatomer epsilon subunit; coatomer protein complex subunit epsilon; coatomer protein complex, subunit epsilon; coatomer subunit epsilon; Cope; Cope1; epsilon coat protein; epsilon-coat protein; Epsilon-COP; nonclathrin coat protein epsilon-COP | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11316 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O14579 | |
| COPE | |
| Recombinant protein corresponding to Human COPE. Recombinant protein control fragment (Product #RP-102912). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction