Learn More
Invitrogen™ COPD Monoclonal Antibody (P189)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA518294
Description
Recommended positive controls: 293T, A431, HeLa, HepG2, Neuro 2A, GL261, C8D30, NIH-3T3, BCL-1, Raw264.7, C2C12, PC-12, Rat2. Predicted reactivity: Mouse (98%), Rat (98%), Xenopus laevis (82%), Chicken (92%), Rhesus Monkey (99%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene maps in a region, which include the mixed lineage leukemia and Friend leukemia virus integration 1 genes, where multiple disease-associated chromosome translocations occur. It is an intracellular protein. Archain sequences are well conserved among eukaryotes and this protein may play a fundamental role in eukaryotic cell biology. It has similarities to heat shock proteins and clathrin-associated proteins, and may be involved in vesicle structure or trafficking.
Specifications
| COPD | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P48444, Q5XJY5, Q66H80 | |
| ARCN1 | |
| Recombinant protein encompassing a sequence within the center region of human COPD. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
| P189 | |
| Unconjugated | |
| ARCN1 | |
| 4632432M07Rik; Archain; archain 1; archain vesicle transport protein 1; Arcn1; coatomer delta subunit; coatomer delta-COP; coatomer protein complex, subunit delta; coatomer protein delta-COP; coatomer subunit delta; COPD; delta-coat protein; Delta-COP; nur17; pale coat neuro | |
| Mouse | |
| Protein G | |
| RUO | |
| 213827, 300674, 372 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.