missing translation for 'onlineSavingsMsg'
Learn More

COPB2 Antibody (M3A5), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 5 publications

£170.00

Specifications

Antigen COPB2
Clone M3A5
Dilution Western Blot, Immunocytochemistry/Immunofluorescence 1:20-1:50, Immunoprecipitation 1:10-1:500, Immunoaffinity Purification
Applications Western Blot
Classification Monoclonal
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18527241
View Documents
Novus Biologicals
NB600-102-0.025ml
0.025 mL
£170.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

COPB2 Monoclonal specifically detects COPB2 in Human, Rat, Avian, Bovine, Canine, Chicken, Hamster, Monkey, Primate, Rabbit samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications

Specifications

COPB2
Western Blot, Immunocytochemistry/Immunofluorescence 1:20-1:50, Immunoprecipitation 1:10-1:500, Immunoaffinity Purification
Monoclonal
Ascites
RUO
Human
9276
Microtubule-associated protein from goose brain.
Primary
Store at -20°C. Avoid freeze/thaw cycles.
M3A5
Western Blot
Unconjugated
Mouse
Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Neuroscience
beta prime subunit, Beta'-coat protein, Beta'-COP, betaprime-COP, coatomer protein complex, subunit beta 2 (beta prime), coatomer subunit beta', p102
COPB2
IgG1
Unpurified
Recognizes an epitope shared by the beta-COP protein (110kD) found in most tissue culture cells, and a high MW doublet of brain microtubule-associated protein (MAP 2, 270-300 kD).
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.