missing translation for 'onlineSavingsMsg'
Learn More

Copanlisib, MedChemExpress

Product Code. 16472567
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16472567 50 mg 50mg
16449057 5 mg 5mg
16436468 10 mg 10mg
16426746 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16472567 Supplier Medchem Express Supplier No. HY15346
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Copanlisib (BAY 80-6946) is a potent, selective and ATP-competitive pan-class I PI3K inhibitor, with IC50s of 0.5 nM, 0.7 nM, 3.7 nM and 6.4 nM for PI3Kα, PI3Kδ, PI3Kβ and PI3Kγ, respectively. Copanlisib has more than 2,000-fold selectivity against other lipid and protein kinases, except for mTOR. Copanlisib has superior antitumor activity.

Copanlisib (BAY 80-6946) is a potent, selective and ATP-competitive pan-class I PI3K inhibitor, with IC50s of 0.5 nM, 0.7 nM, 3.7 nM and 6.4 nM for PI3Kα, PI3Kδ, PI3Kβ and PI3Kγ, respectively. Copanlisib has more than 2,000-fold selectivity against other lipid and protein kinases, except for mTOR. Copanlisib has superior antitumor activity.

Specifications

Chemical Name or Material Copanlisib
CAS 1032568-63-0
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C23H28N8O4
Quantity 50 mg
Synonym BAY 80-6946
Solubility Information 1M HCl : 100 mg/mL (208.11 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (2.08 mM; Need ultrasonic) ∣H2O : 0.67 mg/mL (1.39 mM; Need ultrasonic)
SMILES O=C(C1=CN=C(N)N=C1)NC2=NC3=C(OC)C(OCCCN4CCOCC4)=CC=C3C5=NCCN25
Molecular Weight (g/mol) 480.52
Formula Weight 480.52
Percent Purity 98.02%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.