missing translation for 'onlineSavingsMsg'
Learn More

Copanlisib, MedChemExpress

Product Code. 16436468
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16436468

Brand: Medchem Express HY15346

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Copanlisib (BAY 80-6946) is a potent, selective and ATP-competitive pan-class I PI3K inhibitor, with IC50s of 0.5 nM, 0.7 nM, 3.7 nM and 6.4 nM for PI3Kα, PI3Kδ, PI3Kβ and PI3Kγ, respectively. Copanlisib has more than 2,000-fold selectivity against other lipid and protein kinases, except for mTOR. Copanlisib has superior antitumor activity.

Copanlisib (BAY 80-6946) is a potent, selective and ATP-competitive pan-class I PI3K inhibitor, with IC50s of 0.5 nM, 0.7 nM, 3.7 nM and 6.4 nM for PI3Kα, PI3Kδ, PI3Kβ and PI3Kγ, respectively. Copanlisib has more than 2,000-fold selectivity against other lipid and protein kinases, except for mTOR. Copanlisib has superior antitumor activity.

Specifications

Chemical Name or Material Copanlisib
CAS 1032568-63-0
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C23H28N8O4
Quantity 10 mg
Synonym BAY 80-6946
Solubility Information 1M HCl : 100 mg/mL (208.11 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (2.08 mM; Need ultrasonic) ∣H2O : 0.67 mg/mL (1.39 mM; Need ultrasonic)
SMILES O=C(C1=CN=C(N)N=C1)NC2=NC3=C(OC)C(OCCCN4CCOCC4)=CC=C3C5=NCCN25
Molecular Weight (g/mol) 480.52
Formula Weight 480.52
Percent Purity 98.02%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.