missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
COP9 signalosome complex subunit 2 Polyclonal antibody specifically detects COP9 signalosome complex subunit 2 in Human, Mouse, Rat, Drosophila samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | COP9 signalosome complex subunit 2 |
| Dilution | Western Blot |
| Formulation | PBS |
| Form | Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?