missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568382
This item is not returnable.
View return policy
Description
COP1 Polyclonal Antibody for Western Blot, IHC (P)
Caspase inhibitor. Acts as a regulator of procaspase-1/CASP1 activation implicated in the regulation of the proteolytic maturation of pro-interleukin-1 beta (IL1B) and its release during inflammation. Inhibits the release of IL1B in response to LPS in monocytes. Also induces NF-kappa-B activation during the pro-inflammatory cytokine response. Also able to inhibit CASP1-mediated neuronal cell death, TNF-alpha, hypoxia-, UV-, and staurosporine-mediated cell death but not ER stress-mediated cell death. Acts by preventing activation of caspases CASP1 and CASP4, possibly by preventing the interaction between CASP1 and RIPK2. [UniProt].
Specifications
| COP1 | |
| Polyclonal | |
| Unconjugated | |
| CARD16 | |
| CARD only domain-containing protein 1; CARD only protein; CARD16; CARD-only protein 1; caspase recruitment domain family member 16; caspase recruitment domain family, member 16; caspase recruitment domain-containing protein 16; Caspase recruitment domain-only protein 1; caspase-1 dominant-negative inhibitor pseudo-ICE; caspase-1 inhibitor COP; Constitutive photomorphogenesis protein 1 homolog; constitutive photomorphogenic protein 1; COP; COP1; E3 ubiquitin-protein ligase COP1; E3 ubiquitin-protein ligase RFWD2; hCOP1; Pseudo interleukin-1 beta converting enzyme; pseudo interleukin-1beta converting enzyme; PSEUDO-ICE; pseudo-IL1B-converting enzyme; putative ubiquitin ligase COP1; RFWD2; ring finger and WD repeat domain 2; ring finger and WD repeat domain 2, E3 ubiquitin protein ligase; RING finger and WD repeat domain protein 2; RING finger protein 200; RING-type E3 ubiquitin transferase RFWD2; RNF200 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 114769 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 2 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q5EG05 | |
| CARD16 | |
| Synthetic peptide corresponding to residues near the N terminal of human CARD16. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction