missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COP1 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33503-20ul
This item is not returnable.
View return policy
Description
COP1 Monoclonal antibody specifically detects COP1 in Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| COP1 | |
| Monoclonal | |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| Constitutive photomorphogenesis protein 1 homolog, constitutive photomorphogenic protein (COP1), COP1RNF200FLJ10416, E3 ubiquitin-protein ligase RFWD2, EC 6.3.2.-, hCOP1, putative ubiquitin ligase COP1, ring finger and WD repeat domain 2, RING finger and WD repeat domain protein 2, RING finger protein 200 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 582-731 of human COP1 (NP_071902.2).,, Sequence:, YYDLRNTKQPIMVFKGHRKAVSYAKFVSGEEIVSASTDSQLKLWNVGKPYCLRSFKGHINEKNFVGLASNGDYIACGSENNSLYLYYKGLSKTLLTFKFDTVKSVLDKDRKEDDTNEFVSAVCWRALPDGESNVLIAANSQGTIKVLELV | |
| 20 μL | |
| p53 Pathway, Zinc Finger | |
| 64326 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction