missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COP Antibody (3E10), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00114769-M01
This item is not returnable.
View return policy
Description
COP Monoclonal antibody specifically detects COP in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA
Specifications
| COP | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| CARD only domain-containing protein 1, CARD only protein, caspase recruitment domain family, member 16, Caspase-1 inhibitor COP, COP1caspase-1 dominant-negative inhibitor pseudo-ICE, COPcaspase recruitment domain-containing protein 16, EC 3.4.22.36, Pseudo interleukin-1 beta converting enzyme, pseudo interleukin-1beta converting enzyme, Pseudo-ICE | |
| COPl (NP_443121, 1 a.a. ∽ 97 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MADKVLKEKRKLFIHSMGEGTINGLLDELLQTRVLNQEEMEKVKRENATVMDKTRALIDSVIPKGAQACQICITYICEEDSYLAETLGLSAGPIPGN | |
| 0.1 mg | |
| Apoptosis | |
| 114769 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2b κ |
| Western Blot, ELISA, Sandwich ELISA | |
| 3E10 | |
| Western Blot 1:500, ELISA, Sandwich ELISA | |
| NP_443121 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction