missing translation for 'onlineSavingsMsg'
Learn More

Coomassie Brilliant Blue G-250, MP Biomedicals™

Product Code. 11475951 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
Unit Size:
100g
10g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11475951 100 g 100g
11455951 5 g 5g
11441642 10 g 10g
11465951 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11475951 Supplier MP Biomedicals Supplier No. 0219034380
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Useful dye for SDS gels as it readily stains proteins with minimal background color

  • It is a protein stain in electrophoresis.
  • It has been used in the Bradford dye-binding protein assay.
  • A mechanism for dye binding to protein has been proposed, based on measuring Coomassie Brilliant Blue G-250 (CBBG) absorbance spectra during titration of the dye reagent in the absence of protein and its response to different polyamino acids.
  • Brilliant Blue G has been used for determining critical micelle concentration of detergents.

Chemical Identifiers

CAS 6104-58-1
Molecular Formula C47H48N3NaO7S2
MDL Number MFCD00078482

Specifications

Loss on Drying 0.045
UV_Vis λ max (methanol) 608 ± 3 nm
Beilstein 5230822
Synonym Acid Blue 90, Coomassieg250, CBBG
Color Dark Reddish Purple
Solubility Information Soluble in hot water (50mg/mL), ethanol (40mg/mL), methanol and 10% trichloroacetic acid. Soluble in water (1mg/mL) yielding dark Blue Solution.
Quantity 100 g
InChI Key CMGWKYXJIYYESN-UHFFFAOYSA-M
Physical Form Powder
Molecular Weight (g/mol) 854.02
Grade Not Available
Chemical Name or Material Coomassie Brilliant Blue G-250
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.