missing translation for 'onlineSavingsMsg'
Learn More

Coomassie Brilliant Blue G-250, MP Biomedicals™

Product Code. 11455951 Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
Unit Size:
100g
10g
25g
5g

Product Code. 11455951

Brand: MP Biomedicals 0219034305

Please to purchase this item. Need a web account? Register with us today!

Useful dye for SDS gels as it readily stains proteins with minimal background color

  • It is a protein stain in electrophoresis.
  • It has been used in the Bradford dye-binding protein assay.
  • A mechanism for dye binding to protein has been proposed, based on measuring Coomassie Brilliant Blue G-250 (CBBG) absorbance spectra during titration of the dye reagent in the absence of protein and its response to different polyamino acids.
  • Brilliant Blue G has been used for determining critical micelle concentration of detergents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6104-58-1
Molecular Formula C47H48N3NaO7S2
PubChem CID 6328534
IUPAC Name sodium;3-[[4-[(Z)-[4-(4-ethoxyanilino)phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]-2-methylcyclohexa-2,5-dien-1-ylidene]methyl]-N-ethyl-3-methylanilino]methyl]benzenesulfonate
SMILES CCN(CC1=CC(=CC=C1)S(=O)(=O)[O-])C2=CC(=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC(=CC=C4)S(=O)(=O)[O-])C=C3C)C5=CC=C(C=C5)NC6=CC=C(C=C6)OCC)C.[Na+]

Specifications

Synonym Acid Blue 90, Brilliant Blue G
InChI Key RWVGQQGBQSJDQV-UHFFFAOYSA-M
Molecular Weight (g/mol) 854.025
Formula Weight 854
Grade Electrophoresis
Color Blue to Purple
Quantity 5 g
Physical Form Powder
Chemical Name or Material Coomassie Brilliant Blue G-250, Protein stain in electrophoresis.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.