Learn More
Corning™ CoolRack™ Plate and Reservoir Modules
BIOPHARMA
Product Code. 15552841
Click to view available options
Holds:
1 Flat Bottom Plate, 1 SBS-compatible
1 U-bottom Microplate, SBS compatible
Reagent Reservoir (55 mL)
Unit Size:
Each
Product Code. 15552841
Brand: Corning™ 432070
Only null left
Add to basket
Add to basket
Ensures ±0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
Corning CoolSink thermo-conductive plate and reservoir modules provide uniform temperature to all wells, regardless of position. When placed onto a temperature source such as ice, dry ice, liquid nitrogen or water baths, the CoolSink module will rapidly adapt to that temperature - from -196°C to >+100°C.
  • CoolSink modules ensure temperature uniformity when cooling, snap freezing, heating or thawing samples.
  • All CoolSink modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
  • All modules are compatible with all temperature sources.
  • SBS-compatible
TRUSTED_SUSTAINABILITY
Specifications
Holds 1 Flat Bottom Plate, 1 SBS-compatible
Design CoolSink XT 96F
Type CoolSink Plate and Reservoir Module