missing translation for 'onlineSavingsMsg'
Learn More

Corning™ CoolRack™ M Microcentrifuge Tube Modules
BIOPHARMA

Product Code. 15592801
Change view
Click to view available options
Holds:
15 x 1.5 or 2 mL MCT Tubes
24 x 1.5 or 2 mL MCT Tubes, SBS-compatible
30 x 1.5 or 2 mL MCT Tubes
6 x 1.5 or 2 mL MCT Tubes
90 x 1.5 or 2 mL MCT Tubes
96 x 1.5 or 2 mL MCT Tubes with A to H and 1 to 12
Color:
Gray
Green
Orange
Design:
M15
M24
M30
M6
M90
M96
Unit Size:
Each
Product Code. Holds Color Design unitSize
15592801 6 x 1.5 or 2 mL MCT Tubes Gray M6 Each
15502811 6 x 1.5 or 2 mL MCT Tubes Green M6 Each
15512811 6 x 1.5 or 2 mL MCT Tubes Orange M6 Each
15522811 15 x 1.5 or 2 mL MCT Tubes Gray M15 Each
15532811 15 x 1.5 or 2 mL MCT Tubes Green M15 Each
15542811 15 x 1.5 or 2 mL MCT Tubes Orange M15 Each
15552811 24 x 1.5 or 2 mL MCT Tubes, SBS-compatible Gray M24 Each
15562811 30 x 1.5 or 2 mL MCT Tubes Gray M30 Each
15572811 30 x 1.5 or 2 mL MCT Tubes Green M30 Each
15582811 30 x 1.5 or 2 mL MCT Tubes Orange M30 Each
15592811 90 x 1.5 or 2 mL MCT Tubes Gray M90 Each
15502821 96 x 1.5 or 2 mL MCT Tubes with A to H and 1 to 12 Gray M96 Each
12 options
This item is not returnable. View return policy

Product Code. 15592801

Brand: Corning™ 432034

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Thermo-conductive microfuge tube modules that eliminate variability which originates from tubes placed directly into ice, dry ice, alcohol baths, water baths and other temperature sources.

Corning CoolRack thermo-conductive microcentrifuge tube modules eliminate variability which originates from tubes placed directly into ice, dry ice, alcohol baths, water baths and other temperature sources. Place the CoolRack module directly onto a temperature source between -196°C to >100°C and it will rapidly adapt to that temperature.

  • Ensures +/- 0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
  • All CoolRack modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
  • Suggested applications include cooling reagents such as restriction enzymes, dNTPs and antibodies, alcohol-free dry ice snap freezing of tissue, virus and bacteria samples and bench top cryogenic tube sorting in liquid nitrogen.
TRUSTED_SUSTAINABILITY

Specifications

Color Gray
For Use With (Application) Cooling reagents such as restriction enzymes, dNTPs and antibodies, alcohol-free dry ice snap freezing of tissue, virus and bacteria samples and bench top cryogenic tube sorting in liquid nitrogen
For Use With (Equipment) Microcentrifuge Tubes
Holds 6 x 1.5 or 2 mL MCT Tubes
Array 2 x 3
Design M6
Type Microcentrifuge Tube Modules
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.