missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ CoolRack™ LX50, for use with Axygen 50mL Reagent Reservoir Module

Click to view available options
:
CoolRack™ LX50, for use with Axygen 50mL Reagent Reservoir Module
Description
Corning™ CoolRack™, thermo-conductive reservoir modules provide uniform temperature to all wells, regardless of position. When placed onto a temperature source such as ice, dry ice, liquid nitrogen or water baths, the CoolRack module will rapidly adapt to that temperature - from -196°C to >+100°C.
- CoolRack modules ensure temperature uniformity when cooling, snap freezing, heating or thawing samples.
- All CoolRack modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
- All modules are compatible with all temperature sources.
- For use with the 50 mL Axygen™ disposable reservoirs.
Specifications
Specifications
| For Use With (Equipment) | Axygen 50 mL Reagent Reservoir Module |
| Quantity | 1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction