missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ CoolRack™ 96 Storage Tube Modules

Click to view available options
Holds:
96 x 0.5 mL 2D Bar Code Tubes
96 x 1.4 mL 2D Bar Code Tubes
Unit Size:
Each
Description
Corning CoolRack thermo-conductive tube modules eliminate variability which originates from tubes placed directly into ice, dry ice, alcohol baths, water baths and other temperature sources. Place the CoolRack module directly onto a temperature source between -196°C to >100°C and it will rapidly adapt to that temperature.
- 2D Barcode storage tube modules
- Ensures +/- 0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
- All CoolRack modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
- Suggested applications include cooling reagents such as restriction enzymes, dNTPs and antibodies, alcohol-free dry ice snap freezing of tissue, virus and bacteria samples and bench top cryogenic tube sorting in liquid nitrogen.
- Certain CoolRack modules are SBS-compatible.
Specifications
Specifications
| Holds | 96 x 0.5 mL 2D Bar Code Tubes |
| Type | Storage Tube Module |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction