missing translation for 'onlineSavingsMsg'
Learn More

Corning™ CoolRack™ 96 Storage Tube Modules
BIOPHARMA

Product Code. 15512831
Click to view available options
Holds:
96 x 0.5 mL 2D Bar Code Tubes
96 x 1.4 mL 2D Bar Code Tubes
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15512831

Brand: Corning™ 432056

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Ensures ±0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.

Corning CoolRack thermo-conductive tube modules eliminate variability which originates from tubes placed directly into ice, dry ice, alcohol baths, water baths and other temperature sources. Place the CoolRack module directly onto a temperature source between -196°C to >100°C and it will rapidly adapt to that temperature.

  • 2D Barcode storage tube modules
  • Ensures +/- 0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
  • All CoolRack modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
  • Suggested applications include cooling reagents such as restriction enzymes, dNTPs and antibodies, alcohol-free dry ice snap freezing of tissue, virus and bacteria samples and bench top cryogenic tube sorting in liquid nitrogen.
  • Certain CoolRack modules are SBS-compatible.
TRUSTED_SUSTAINABILITY

Specifications

Holds 96 x 0.5 mL 2D Bar Code Tubes
Type Storage Tube Module
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.