missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP BIOMEDICALS CoolPrep Adapter Bottom Component
Shop All MP Biomedicals Products
Click to view available options
For Use With (Equipment):
FastPrep-24
Unit Size:
Each
Description
- Replacement bottom component of CoolPrep adapter.
- Holds 24 x 2 mL tubes.
Usage Statement:
- Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Specifications
Specifications
| Product Type | CoolPrep™ Adapter |
| For Use With (Equipment) | FastPrep-24 |
| Holds | 24 Tubes (2 mL) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction