missing translation for 'onlineSavingsMsg'
Learn More

LMS™ Series 4 Model 1200 Cooled Incubator, 1200 L, Stainless Steel

Product Code. 11887602
Change view
Click to view available options
Capacity (Metric):
1200 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
11887602 1200 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11887602 Supplier LMS™ Supplier No. 1200

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Temperature Range: -10°C to +50°C

  • Reliable and accurate temperature control
  • Temperature range -10.0°C to +50.0°C
  • Models within this series include 600L and 1200L capacity incubators as either single or dual temperature (24 hour or 7 day cycle) versions, or a fully programmable 1200L capacity incubator providing multiple temperature control
  • Stainless steel exterior and interior construction with foamed polyurethane insulation
  • Energy efficient and eco-friendly
  • P.I.D. heating and cooling to ensure stable internal temperature
  • Pre-set safety cut-out
  • Hermetically sealed refrigeration system
  • Door lock
  • Controls easy to read and simple to use
  • Temperature variation with average workload ±0.5°C
  • Full range of factory-fitted optional extras available, including LED lighting, access porthole, inner perspex doors, outer glazed doors, alarm system, internal socket and chart recorder
  • Ideal for applications such as growth studies, B.O.D.tests, seed germination and vernalisation, tissue culture, enzymology, serum studies and immunology, sample storage or product shelf life tests

Specifications

Certifications/Compliance ISO 9001:2008
Controller Type Standard digital
Description Freestanding incubator
Capacity (Metric) 1200 L
Chamber Material Stainless Steel
Convection Type Fan-assisted Circulation
Dimensions (D x W) Shelf 550 mm x 530 mm
Lighting Optional LED
Material Stainless Steel
No. of Shelves 10
Temperature Range (Metric) -10°C to +50°C
Type Refrigerated Incubator
Display Digital
Insulation Foamed polyurethane
Model 1200
No. of Doors 2
Refrigerant CFC-free
Temperature Stability ±0.1°C
Temperature Uniformity ±0.5°C
Warranty 2 Years
Width (English) 59.0 in.
Width (English) Interior 53.1 in.
Width (Metric) 1500 mm
Width (Metric) Interior 1350 mm
Height (English) 81.9 in.
Height (English) Interior 59.0 in.
Height (Metric) 2080 mm
Height (Metric) Interior 1500 mm
Depth (English) Interior 26.4 in.
Depth (Metric) Interior 670 mm
Weight (Metric) 235 kg
Electrical Requirements 240 V, 50 Hz
Frequency 50 Hz
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.