missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ CoolBox™ XT Systems

Click to view available options
Color:
Green
Orange
Pink
Purple
Includes:
CoolBox 2XT System, CoolRack PCR96, SBS-Compatible CoolRack XT M24
CoolBox XT System, SBS-Compatible CoolRack XT M-PCR
CoolBox XT System, SBS-Compatible CoolRack XT M24
Holder and One Cooling Core
Holder and Two Cooling Cores
Product Line:
CoolBox 2XT
CoolBox 2XT PCR system
CoolBox XT
CoolBox XT M24 system
CoolBox XT PCR Strip system
Unit Size:
Each
Description
Corning CoolBox XT systems are bench top cooling workstations that provide sample cooling or freezing without ice, electricity or batteries. CoolBox systems are versatile and accommodate a variety of sample formats and temperatures. The modular design enables the use of CoolRack™ and CoolSink™ thermo-conductive sample modules to hold microfuge tubes, cryogenic vials, PCR tubes or plates, assay plates and more.
- Consists of a base, cooling cartridge/core and a lid
- Includes a magnetized collar and a 1° to 8°C temperature indicator for visual assurance of proper temperature
- To use with any 30 place Corning CoolRack Tubes Module
- The internal cooling or freezing cores provide 0.5° to 4°C for over 10 hours with lid open and over 16 hours with lid closed
- CoolBox systems may also be used with dry ice to create a compact, portable, snap freezing workstation for bacteria, virus or proteins.
Spécification
Spécification
| Description | Benchtop cooling system |
| Color | Purple |
| Type | Cool Box |
| For Use With (Application) | Enables Cooling for 16 Hours, Freezing for 8 Hours |
| Holds | 30 Place |
| Includes | Holder and One Cooling Core |
| Product Line | CoolBox XT |
| Weight (Metric) | 3 kg |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu