Learn More
Epredia™ Cool-Cut with universal cassette clamp
Maintain a cool, consistent temperature for paraffin blocks during the sectioning process with the Epredia™ Cool-Cut accessory.
Brand:  Epredia™ 771110
Code : 50
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 11336770
  • £1905.00 / Each
Estimated Shipment: 21-06-2024
to see stock.
Description
Description
  • Directly fits onto the specimen cylinder of the rotary microtome line
  • Ability to be equipped as a universal cassette clamp or standard specimen clamp
  • Effective when extended cutting periods on the microtome are required to “step section” or “serial section” through the paraffin block
Includes:
  • Peltier device with integrated specimen clamp
  • Counter-balance weights
  • Power supply
Specifications
Specifications
With universal cassette clamp
Microtome Accessory
Epredia Microtomes